Learn More
Aviva Systems Biology Corp CEACAM3 Antibody - C-terminal region (ARP60120_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60120_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEACAM3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CEACAM3. Alternate names: CD66D, CEA, CGM1, MGC119875, W264, W282ApplicationWB. Species reactivity: Human
Catalog No. 50-286-947