Learn More
Sino Biological CEACAM5 / CD66e Antibody (APC), Rabbit MAb 25Tests
Supplier:  Sino Biological 11077R061A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM5 / CD66e (rh CEACAM5 / CD66e; Catalog#11077-H08H; NP_004354.2; Met1-Ala685) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-224-7466