Learn More
Sino Biological CEACAM5 / CD66e Antibody (PE), Mouse MAb 100Tests
Supplier:  Sino Biological 11077MM02P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM5 / CD66e (rh CEACAM5 / CD66e; Catalog#11077-H08H; NP_004354.2; Met1-Ala685) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-7425