Learn More
ACROBiosystems Recombinant Protein;Human CEACAM-5 / CD66e Protein;HEK293;1MG;CE5,CEACAM-5,CD66e,CEACAM5,CEA,Meconium antigen 100
Supplier:  ACROBiosystems CE5H52261MG
Encompass_Preferred
Recombinant Protein;Human CEACAM-5 / CD66e Protein;1MG;Human CEACAM-5 (CE5-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.2 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7531