Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CEACAM-5 / CD66e Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;CE5,CEACAM-5,CD66e,CEACAM5,CEA,Meconium antigen 100
Supplier:  ACROBiosystems CE5H82E0200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CEACAM-5 / CD66e Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CEACAM-5, His Tag, Avi Tag (CE5-H82E0) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Ala 685 (Accession # P06731-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 75.0 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7488