Learn More
Aviva Systems Biology Corp CEACAM6 Antibody - middle region (ARP41504_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41504_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CEACAM6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CEACAM6. Alternate names: CD66c, CEAL, NCAApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57885