Learn More
Enzo Life Sciences CEACAM6 (human) monoclonal antibody (9A6) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805083C100
Encompass_Preferred
Clone: 9A6. Isotype: IgG1. Immunogen: Tumor cell lines expressing human CEACAM6. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC (FS), WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2. Contains no preservatives.
Catalog No. 50-201-0054