Learn More
Aviva Systems Biology Corp CEACAM7 Antibody - C-terminal region (ARP60123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEACAM7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CEACAM7. Alternate names: CEA, CGM2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-286-949