Learn More
Aviva Systems Biology Corp CEACAM7 Antibody - N-terminal region (ARP60122_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60122_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEACAM7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CEACAM7. Alternate names: CEA, CGM2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-948