Learn More
ACROBiosystems Recombinant Protein;Human CEACAM-8 / CD66b Protein;HEK293;100UG;CE8,CEACAM8,CEACAM-8,CEACAM 8,CD66b,CD67,CGM6,NCA-95,NCA95,NCA 95
Catalog No. 501657518
Encompass_Preferred
Recombinant Protein;Human CEACAM-8 / CD66b Protein;100UG;Human CEACAM-8, His Tag (CE8-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession # NP_001807.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33 kDa. The protein migrates as 48-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7518 Supplier ACROBiosystems Supplier No. CE8H5224100UG
Recombinant Protein;Human CEACAM-8 / CD66b Protein;100UG;Human CEACAM-8, His Tag (CE8-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession # NP_001807.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33 kDa. The protein migrates as 48-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.