Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CEACAM-8 / CD66b Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;CE8,CEACAM8,CEACAM-8,CEACAM 8,CD66b,CD67,CGM6,NCA-95,NCA95,NCA 95
Supplier:  ACROBiosystems CE8H82E9200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CEACAM-8 / CD66b Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CEACAM-8, His Tag, Avi Tag (CE8-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession # P31997-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 35.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CEACAM-6, His Tag (Cat. No. CE6-H5223) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-8, His Tag, Avi Tag (Cat. No. CE8-H82E9) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-165-7880