Learn More
TARGETMOL CHEMICALS INC CEAPIN-A7 25MG
SDP
Catalog No. 502579020
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ceapin-A7 is a selective blocker of ATF6(alpha) signaling in response to ER stress with an IC50 of 0.59 uM. Ceapin-A7 can be used to explore both the mechanism of activation of ATF6(alpha) and its role in pathological settings. purity: 99%
Catalog No. 50-257-9020 Supplier TARGETMOL CHEMICALS INC Supplier No. T911025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ceapin-A7 is a selective blocker of ATF6(alpha) signaling in response to ER stress with an IC50 of 0.59 uM. Ceapin-A7 can be used to explore both the mechanism of activation of ATF6(alpha) and its role in pathological settings. purity: 99%