Learn More
Selleck Chemical LLC Ceapin-A7-E1099-5MG
SDP
Supplier:  Selleck Chemical LLC E10995MG
Encompass_Preferred
Ceapin-A7 is a selective and highly potent inhibitor of the ATF6 which can activate the response to ER stress with IC50 of 0 59 M
Catalog No. 50-238-9807