Learn More
Medchemexpress LLC HY-108434 5mg Medchemexpress, Ceapin-A7 CAS:2323027-38-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084345MG
Encompass_Preferred
Medchemexpress, HY-108434 5mg Ceapin-A7 CAS:2323027-38-7 Ceapin-A7 is a selective blocker of ATF6α signaling in response to ER stress, with an IC50 of 0.59 μM. Ceapin-A7 can be used to explore both the mechanism of activation of ATF6α and its role in pathological settings. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0743