Learn More
Medchemexpress LLC CEBP gamma/CEBPG Hu 10ug
SDP
Catalog No. 5000178389
Encompass_Preferred
CEBP gamma/CEBPG protein is a transcription factor that regulates genes by binding to their promoter and enhancer regions It targets IL-4 and immunoglobulin heavy chains binding to enhancer elements PRE-I and GPE1 in the G-CSF gene promoter CEBP gamma/CEBPG Protein Human (His) is the recombinant human-derived CEBP gamma/CEBPG protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-78389 Supplier Medchemexpress LLC Supplier No. HYP7425410UG
CEBP gamma/CEBPG protein is a transcription factor that regulates genes by binding to their promoter and enhancer regions It targets IL-4 and immunoglobulin heavy chains binding to enhancer elements PRE-I and GPE1 in the G-CSF gene promoter CEBP gamma/CEBPG Protein Human (His) is the recombinant human-derived CEBP gamma/CEBPG protein expressed by E coli with N-His labeled tag