Learn More
Aviva Systems Biology Corp CEBPA Antibody - C-terminal region (ARP57938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEBPA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CEBPA. Alternate names: C/EBP-alpha, CEBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-416