Learn More
Aviva Systems Biology Corp CEBPA Antibody - middle region (ARP57937_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57937_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEBPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CEBPA. Alternate names: C/EBP-alpha, CEBPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62182