Learn More
Aviva Systems Biology Corp CEBPA Antibody - N-terminal region (P100908_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100908_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEBPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CEBPA. Alternate names: CEBP, C/EBP-alphaApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-53804