Learn More
Medchemexpress LLC Recombinant human CEBPA protein (His-tag) | >95.0% | 10UG
SDP
Catalog No. 5000174851
Encompass_Preferred
Recombinant human CEBPA is a His-tagged transcription factor produced in Escherichia coli and supplied lyophilized for research applications. The product is suitable for in vitro biochemical assays, antibody validation, and studies of transcriptional regulation.
  • Recombinant human CEBPA protein with N-terminal His tag.
  • Expressed in Escherichia coli and purified to >95.0% purity.
  • Approximate molecular weight 44 kDa.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • Available in small sizes such as 10 μg for micro-scale experiments.
Catalog No. 50-001-74851 Supplier Medchemexpress LLC Supplier No. HYP70122710UG
Recombinant human CEBPA is a His-tagged transcription factor produced in Escherichia coli and supplied lyophilized for research applications. The product is suitable for in vitro biochemical assays, antibody validation, and studies of transcriptional regulation.
  • Recombinant human CEBPA protein with N-terminal His tag.
  • Expressed in Escherichia coli and purified to >95.0% purity.
  • Approximate molecular weight 44 kDa.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • Available in small sizes such as 10 μg for micro-scale experiments.