Learn More
Aviva Systems Biology Corp ADA2 Antibody - N-terminal region (ARP47298_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47298_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CECR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CECR1. Alternate names: IDGFL, ADA2, ADGFApplicationWB. Species reactivity: Human
Catalog No. 50-014-59512