Learn More
TARGETMOL CHEMICALS INC Cecropin A 1MG
SDP
Catalog No. 502879422
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cecropin A, a linear 37-residue antimicrobial polypeptide produced by the cecropia moth, has exhibited cytotoxicity in various human cancer cell lines and inhibitory effects on tumor growth. Purity 96.78%
Catalog No. 50-287-9422 Supplier TARGETMOL CHEMICALS INC Supplier No. TP14101MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cecropin A, a linear 37-residue antimicrobial polypeptide produced by the cecropia moth, has exhibited cytotoxicity in various human cancer cell lines and inhibitory effects on tumor growth. Purity 96.78%