Learn More
Cecropin A, G-Biosciences

Description
Cecropin A produces synergistic effects with some chemotherapeutic agents, enhancing cytotoxic activity. Cecropin A has been shown to cause bacterial and cancerous cell death by insertion into the cellular membrane due to amphiphilic N-terminal residues.
Specifications
Specifications
| Quantity | 1 mg |
| Percent Purity | >95% |
| Grade | Laboratory |
| Physical Form | Powder |
| Product Line | Gibco |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.