Learn More
Cayman Chemical CecropIn AtrIfluoroacetAT 10mg
SDP
Catalog No. 502915189
Encompass_Preferred
An antimicrobial peptide; induces apoptosis in C. albicans at 5 µM; active against the E. coli strain CFT073 (MIC = 100 UG/ml); increases survival of CFT073-infected G. mellonella larvae; decreases enrichment of the gut microflora Bacteroidaceae and Enterobacteriaceae and increases gut levels of Lactobacillus, as well as increases survival and decreases body weight loss in a mouse model of DSS-induced IBD
Catalog No. 50-291-5189 Supplier Cayman Chemical Supplier No. 3690110 MG
An antimicrobial peptide; induces apoptosis in C. albicans at 5 µM; active against the E. coli strain CFT073 (MIC = 100 UG/ml); increases survival of CFT073-infected G. mellonella larvae; decreases enrichment of the gut microflora Bacteroidaceae and Enterobacteriaceae and increases gut levels of Lactobacillus, as well as increases survival and decreases body weight loss in a mouse model of DSS-induced IBD