Learn More
Cecropin B, G-Biosciences

Description
Cecropin B has been shown to produce in vivo cytotoxic effects against stomach carcinomas. It has also been used to synthesize derivatives with activity against other cancer cell lines.
Specifications
Specifications
| Quantity | 1 mg |
| Percent Purity | >95% |
| Grade | Laboratory |
| Physical Form | Powder |
| Product Line | Gibco |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.