Learn More
Bachem Cecropin P1 Trifluoroacetate
SDP
Supplier:  Bachem 4039862.1000
Encompass_Preferred
Cecropin P1 Trifluoroacetate, H-Ser-Trp-Leu-Ser-Lys-Thr-Ala-Lys-Lys-Leu-Glu-Asn-Ser-Ala-Lys-Lys-Arg-Ile-Ser-Glu-Gly-Ile-Ala-Ile-Ala-Ile-Gln-Gly-Gly-Pro-Arg-OH, 125667-96-1, C147H253N45O43, Mr 3338.90, 1mg. Cecropin P1, porcine. Cecropin P1 (SWLSKTAKKLENSAKKRISEGIAIAIQGGPR), a homolog of insect cecropins isolated from pig intestine, was thought to be the first mammalian antimicrobial peptide. Later studies showed that the peptide is produced by the intestinal parasitic nematode Ascaris suum. The 31 amino acid peptide is highly potent against Gram-negative bacteria but has a reduced activity against Gram-positive bacteria. A disruptive effect on bacterial membranes has been proposed as mechanism of antimicrobial activity.
Catalog No. 50-194-6656