Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cedar Leaf Oil, FCC, 60%, Spectrum™ Chemical
SDP

Catalog No. 18601516 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
125 mL
500 mL
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-516 25 mL Amber Glass Bottle
18-601-517 125 mL Amber Glass Bottle
18-601-518 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-516 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C110925ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1109, 8007-20-3

Spectrum™ Chemical Cedar Leaf Oil, FCC is considered generally safe by the FDA as a food additive preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8007-20-3

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.456-1.460
Quantity 25 mL
UN Number UN1169
Percent Purity 60%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.