Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cedar oil | 8000-27-9 | MFCD00132766 | 25 G
SDP

Catalog No. 5000283288
Encompass_Preferred

Cedar oil is a pure cedarwood essential oil intended for laboratory and analytical use. It is a natural mixture used as a co-solvent, reagent, and reference material in biochemical, formulation, and microscopy applications. Product documentation includes batch-specific COA and SDS files for quality verification.

  • Natural essential oil (cedarwood).
  • CAS number 8000-27-9.
  • Supplied in pure form.
  • Available sizes: 25 G, 50 G, 100 G, 500 G, 1 KG.
  • Solubility: DMSO 200 mg/mL; ethanol 100 mg/mL (ultrasonic recommended).
  • Storage: -20°C for 3 years, 4°C for 2 years.
  • Batch-specific COA and SDS available.

Catalog No. 50-002-83288 Supplier Medchemexpress LLC Supplier No. HYW13389425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cedar oil is a pure cedarwood essential oil intended for laboratory and analytical use. It is a natural mixture used as a co-solvent, reagent, and reference material in biochemical, formulation, and microscopy applications. Product documentation includes batch-specific COA and SDS files for quality verification.

  • Natural essential oil (cedarwood).
  • CAS number 8000-27-9.
  • Supplied in pure form.
  • Available sizes: 25 G, 50 G, 100 G, 500 G, 1 KG.
  • Solubility: DMSO 200 mg/mL; ethanol 100 mg/mL (ultrasonic recommended).
  • Storage: -20°C for 3 years, 4°C for 2 years.
  • Batch-specific COA and SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.