Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cedar oil (oils, cedarwood) | 8000-27-9 | MFCD00132766 | 100 G
SDP

Catalog No. 5000283510
Encompass_Preferred

Cedar oil is an essential cedarwood oil supplied for laboratory and research use as a natural mixture of terpenoid components. The product page and batch documentation provide handling, solubility guidance, storage recommendations, and downloadable technical files (COA, SDS, data sheet) for analytical and formulation work.

  • Essential oil form composed of a natural mixture of cedarwood constituents.
  • Available in multiple sizes, including 100 G packaging.
  • Solubility guidance: DMSO 200 mg/mL (requires ultrasonic), ethanol 100 mg/mL (requires ultrasonic).
  • Recommended storage: pure form -20 °C (3 years) or 4 °C (2 years).
  • Documentation available: certificate of analysis, safety data sheet, and data sheet.
  • Suitable for analytical reference, formulation, and research applications.

Catalog No. 50-002-83510 Supplier Medchemexpress LLC Supplier No. HYW133894100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cedar oil is an essential cedarwood oil supplied for laboratory and research use as a natural mixture of terpenoid components. The product page and batch documentation provide handling, solubility guidance, storage recommendations, and downloadable technical files (COA, SDS, data sheet) for analytical and formulation work.

  • Essential oil form composed of a natural mixture of cedarwood constituents.
  • Available in multiple sizes, including 100 G packaging.
  • Solubility guidance: DMSO 200 mg/mL (requires ultrasonic), ethanol 100 mg/mL (requires ultrasonic).
  • Recommended storage: pure form -20 °C (3 years) or 4 °C (2 years).
  • Documentation available: certificate of analysis, safety data sheet, and data sheet.
  • Suitable for analytical reference, formulation, and research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.