Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cedar Oil, TCI America™
SDP

Catalog No. C062625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C062625ML Supplier TCI America Supplier No. C062625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8000-27-9
MDL Number MFCD00132766
Synonym Cedarwood Oil

Specifications

Physical Form Clear Liquid at 20°C
Quantity 25 mL
Chemical Name or Material Cedar Oil
RTECSNumber FJ1520050
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.