Learn More
Cederlane CEDARLANE LYMPHOLYTE 5X30ML
SDP
Catalog No. NC0221185
Encompass
Lympholyte�-Rat Cell Separation Media (Isolation of lymphocytes from Rat lymphoid tissue), sterile liquid, Size: 5x30ml
Catalog No. NC0221185 Supplier Cederlane Supplier No. CL5040
Lympholyte�-Rat Cell Separation Media (Isolation of lymphocytes from Rat lymphoid tissue), sterile liquid, Size: 5x30ml