Learn More
Medchemexpress LLC Cedarwood oil | 8000-27-9 | MFCD00146327 | 1 KG
SDP
Catalog No. 5000283318
Encompass_Preferred
Cedarwood oil is a natural essential oil distilled from the heartwood of various conifer species. This research-grade material is a complex mixture of terpenes and sesquiterpenes and is used in laboratory studies for its insecticidal, preservative, and aromatic properties. The product is supplied as a liquid; consult the COA or SDS for batch-specific composition and purity.
  • Natural mixture of terpenes and sesquiterpenes.
  • Exhibits pesticidal and preservative properties.
  • Supplied as a colorless to light yellow liquid.
  • Available in packaging up to 1 kg.
  • Consult COA or SDS for batch-specific composition and purity.
  • Intended for research use only; not for patient use.
Catalog No. 50-002-83318 Supplier Medchemexpress LLC Supplier No. HYW1338941KG
Cedarwood oil is a natural essential oil distilled from the heartwood of various conifer species. This research-grade material is a complex mixture of terpenes and sesquiterpenes and is used in laboratory studies for its insecticidal, preservative, and aromatic properties. The product is supplied as a liquid; consult the COA or SDS for batch-specific composition and purity.
  • Natural mixture of terpenes and sesquiterpenes.
  • Exhibits pesticidal and preservative properties.
  • Supplied as a colorless to light yellow liquid.
  • Available in packaging up to 1 kg.
  • Consult COA or SDS for batch-specific composition and purity.
  • Intended for research use only; not for patient use.