Learn More
Cayman Chemical CedazurIdIn 10mg
SDP
Catalog No. 502917200
Encompass_Preferred
A cytidine deaminase inhibitor (IC50 = 0.4 µM); increases plasma levels of decitabine in rhesus monkeys at 1 and 10 mg/kg; induces decreases in leukemic cell expansion in combination with orally administered 5-azacytidine, which is inactive as a single agent, in the bone marrow and spleen in a MOLM-13 AML mouse xenograft model at 3 mg/kg
Catalog No. 50-291-7200 Supplier Cayman Chemical Supplier No. 3876710 MG
A cytidine deaminase inhibitor (IC50 = 0.4 µM); increases plasma levels of decitabine in rhesus monkeys at 1 and 10 mg/kg; induces decreases in leukemic cell expansion in combination with orally administered 5-azacytidine, which is inactive as a single agent, in the bone marrow and spleen in a MOLM-13 AML mouse xenograft model at 3 mg/kg