Learn More
Roche Diagnostics CEDEX BIO ISE DEPROT 2X11ML
Supplier:  Roche Diagnostics 06440517001
Encompass
This product is used to clean residual protein from the sampling probe and ISE electrodes.
Catalog No. 50-589-392