Learn More
Roche Diagnostics CEDEX DETERGENT 1000 ML
Supplier:  Roche Diagnostics 05650496001
Encompass
Cedex Detergent is required for cleaning the capillary system of the Cedex and Cedex HiRes Systems. The ready-to-use solution is available as a component of the Cedex Reagent Kit and as a separate bottle.
Catalog No. 50-589-343