Learn More
Roche Diagnostics Sodium Bio HT, 200 tests
Supplier:  Roche Diagnostics 08881871001
Encompass
Kit for determination of sodium. This product is intended for quantitative determination of sodium in aqueous solutions using the Cedex Bio HT Analyzer.
Catalog No. NC1926119