Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Control Set
GSA_VA

Catalog No. 23278215 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-215 2 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-215 Supplier Thermo Scientific™ Supplier No. 100246
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get world-renowned accuracy, dependability and stability with the Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Control Set, which is optimized for most analyzers.

Get world-renowned accuracy, dependability and stability with the Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Control Set, which is optimized for most analyzers. CEDIA assays offer one of the most extensive menus on the market for drugs of abuse testing.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description CEDIA Buprenorphine Control Set: Low 3 and High 7
Concentration 5 ng/mL, Low (3ng/mL), High (7ng/mL)
Detectable Analytes Buprenorphine
Sample Type Urine
Type Control
Level High, Low
Quantity 2 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.