Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEDIA™ Buprenorphine II Drugs of Abuse Assay

Catalog No. 21200428
Change view
Click to view available options
Quantity:
1 x 65 mL, MCC
3 x 17 mL, TSC
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-428 3 x 17 mL, TSC
21-200-429 1 x 65 mL, MCC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-428 Supplier Thermo Scientific Supplier No. 10020849
Add to Cart
Add to Cart
This item is not returnable. View return policy

Determine the presence of Buprenorphine and all major metabolites with the new, improved formulation of this immunoassay.

Determine the presence of Buprenorphine and its metabolites with the new, improved Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Assay. It is the only assay currently available on the market which detects all three major metabolites: norbuprenorphine, buprenorphine-3-β-glucuronide and norbuprenorphine-3-β-glucuronide. Buprenorphine is widely used as a practical alternative to methadone in the treatment of opioid addiction. This assay has no significant cross-reactivity to other opioids and does not cross-react with morphine, even at 100,000ng/mL.

  • Detect Buprenorphine and its metabolites: buprenorphine-3-β-glucuronide, norbuprenorphine, and norbuprenorphine-3-β-glucuronide
  • Qualitative or semi-quantitative options for results
  • No cross-reactivity with any other opioids
  • Applications available on a wide range of clinical chemistry analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets CEDIA Buprenorphine II Control Set
Description CEDIA Buprenorphine II Assay
Detectable Analytes Buprenorphine (BUP), Norbuprenorphine (NBUP), Buprenorphine-3-β-glucuronide, Norbuprenorphine-3-β-glucuronide
DoA Calibrators Buprenorphine II Calibrator, Negative Calibrator
Sample Type Urine
Type Reagent
Quantity 3 x 17 mL, TSC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.