Learn More
Description
Determine the presence of Buprenorphine and its metabolites with the new, improved Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Assay. It is the only assay currently available on the market which detects all three major metabolites: norbuprenorphine, buprenorphine-3-β-glucuronide and norbuprenorphine-3-β-glucuronide. Buprenorphine is widely used as a practical alternative to methadone in the treatment of opioid addiction. This assay has no significant cross-reactivity to other opioids and does not cross-react with morphine, even at 100,000ng/mL.
- Detect Buprenorphine and its metabolites: buprenorphine-3-β-glucuronide, norbuprenorphine, and norbuprenorphine-3-β-glucuronide
- Qualitative or semi-quantitative options for results
- No cross-reactivity with any other opioids
- Applications available on a wide range of clinical chemistry analyzers
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | CEDIA Buprenorphine II Control Set |
| Description | CEDIA Buprenorphine II Assay |
| Detectable Analytes | Buprenorphine (BUP), Norbuprenorphine (NBUP), Buprenorphine-3-β-glucuronide, Norbuprenorphine-3-β-glucuronide |
| DoA Calibrators | Buprenorphine II Calibrator, Negative Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 65 mL, MCC |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.