Learn More
CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
Description
Achieve qualitative and semi-quantitative test results with the Thermo Scientific™ CEDIA™ Buprenorphine II Calibrators and Controls. The five calibrators, including negative, are used to establish a standard curve that correlates the reaction signals from the analyzer with the calibrators’ respective concentrations (in ng/mL). Two controls, one above and one below the cutoff (10ng/mL), validate the calibration curve. The calibrators and controls also provide a reference point for qualitative (positive or negative results) and semi-quantitative detection.
- Ensures assay accuracy
- 18 months shelf life
- Supports qualitative and semi-quantitative measurements
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | CEDIA Buprenorphine II 10ng/mL Cutoff Calibrator |
| Concentration | 10 ng/mL |
| Detectable Analytes | Buprenorphine |
| Sample Type | Urine |
| Type | Calibrator |
| Quantity | 1 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.