Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
GSA_VA

Catalog No. p-6160001
Change view
Click to view available options
Type:
CsA Control High Level 4
CsA Control High Level 5
CsA PLUS Combo Kit
CsA PLUS High Calibrator Set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-278-294 CsA PLUS Combo Kit
23-278-289 CsA PLUS High Calibrator Set
23-278-291 CsA Control High Level 4
23-278-292 CsA Control High Level 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-278-294 Supplier Thermo Scientific™ Supplier No. 100147
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Achieve optimal low-end sensitivity and a broad dynamic range with the Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.

Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.

Accurate:
  • Excellent low-end sensitivity
  • Correlation with gold standard methods
  • Precise throughput standard curve assay range
  • Lot-to-lot consistency

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Cyclosporin A
Sample Type Plasma, Serum, Plasma, Serum
Type CsA PLUS Combo Kit
Quantity 41 mL, 19 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.