Learn More
Thermo Scientific™ CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays
Shop All Thermo Scientific ProductsDescription
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | MGC Select DAU Control Set |
| Description | CEDIA Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assay for Indiko |
| Detectable Analytes | 6-Acetylmorphine, Heroin Metabolite (6-Acetylmorphine) |
| DoA Calibrators | Heroin Metabolite Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 3 x 17 mL, Indiko |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.