Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays

Catalog No. 23278285 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 495 mL, LC
1 x 65 mL, MCC
3 x 17 mL, Indiko
3 x 17 mL, TSC
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-285 3 x 17 mL, Indiko
23-278-280 3 x 17 mL, TSC
23-278-281 1 x 65 mL, MCC
21-200-436 1 x 495 mL, LC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-278-285 Supplier Thermo Scientific™ Supplier No. 10016433
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA THC (Cannabinoid) Drugs of Abuse Assays.

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.

  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets CEDIA THC Controls
Description CEDIA THC (Cannabinoid) Drugs of Abuse Assay for Indiko
Detectable Analytes 11-nor-Δ9THC-COOH
DoA Calibrators Multi-level THC Calibrator
Sample Type Urine
Type Reagent
Quantity 3 x 17 mL, Indiko
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.