Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators

Catalog No. 23278238
Change view
Click to view available options
Description:
CEDIA Antibiotic TDM Multi-Calibrators
CEDIA Antiepileptic TDM Multi-Calibrators
CEDIA Cardiac TDM Multi-Calibrators
Detectable Analytes:
Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
Gentamicin, Tobramycin
NAPA, Procainamide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Detectable Analytes
23-278-238 CEDIA Cardiac TDM Multi-Calibrators NAPA, Procainamide
23-278-239 CEDIA Antiepileptic TDM Multi-Calibrators Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid
23-278-240 CEDIA Antibiotic TDM Multi-Calibrators Gentamicin, Tobramycin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-278-238 Supplier Thermo Scientific™ Supplier No. 100001
Add to Cart
Add to Cart

Save time while maintaining accuracy with economical Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, optimized for use with CEDIA TDM Assays.

Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.

  • Proven technology
  • Liquid ready-to-use: no reconstitution needed
  • Multiple drug constituents per calibration kit
  • Positive linear calibration curve: the higher the absorbance reading, the more drug present
  • Excellent correlation to reference methods
  • Recombinant DNA technology provides lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description CEDIA Cardiac TDM Multi-Calibrators
Detectable Analytes NAPA, Procainamide
Sample Type Plasma, Serum
Type Calibrator
Quantity Low: 2 x 7.5 mL; High: 2 x 5.0 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.