Learn More
Selleck Chemical LLC Cediranib S1017-5MG
SDP
Catalog No. 5000042606
Encompass_Preferred
Cediranib (AZD2171 NSC-732208) is a highly potent VEGFR(KDR) inhibitor with IC50 of 1 nM also inhibits Flt1/4 with IC50 of 5 nM/ 3 nM similar activity against c-Kit and PDGFR 36- 110-fold and 1000-fold selective more for VEGFR than PDGFR- CSF-1R and Flt3 in HUVEC cells Cediranib (AZD2171) induces autophagic vacuole accumulation Phase 3
Catalog No. 50-000-42606 Supplier Selleck Chemical LLC Supplier No. S10175MG
Cediranib (AZD2171 NSC-732208) is a highly potent VEGFR(KDR) inhibitor with IC50 of 1 nM also inhibits Flt1/4 with IC50 of 5 nM/ 3 nM similar activity against c-Kit and PDGFR 36- 110-fold and 1000-fold selective more for VEGFR than PDGFR- CSF-1R and Flt3 in HUVEC cells Cediranib (AZD2171) induces autophagic vacuole accumulation Phase 3