Learn More
TARGETMOL CHEMICALS INC CEDIRANIB 25MG
SDP
Catalog No. 502577105
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cediranib (AZD2171) (AZD2171) is a highly potent (IC50 < 1 nmol/L) ATP-competitive inhibitor of recombinant KDR tyrosine kinase in vitro also inhibits Flt1/4 (IC50 5 nM/≤3 nM) similar activity against PDGFR(beta) and c-Kit selective more for VEGFR than PDGFR-(alpha) (36-fold) CSF-1R (110-fold) and Flt3 (1000-fold) in HUVEC cells. purity: 99%
Catalog No. 50-257-7105 Supplier TARGETMOL CHEMICALS INC Supplier No. T250025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cediranib (AZD2171) (AZD2171) is a highly potent (IC50 < 1 nmol/L) ATP-competitive inhibitor of recombinant KDR tyrosine kinase in vitro also inhibits Flt1/4 (IC50 5 nM/≤3 nM) similar activity against PDGFR(beta) and c-Kit selective more for VEGFR than PDGFR-(alpha) (36-fold) CSF-1R (110-fold) and Flt3 (1000-fold) in HUVEC cells. purity: 99%