Learn More
Medchemexpress LLC HY-10205 200mg , Cediranib AZD2171 CAS:288383-20-0 Purity:98%
SDP
Catalog No. 501492577
Encompass_Preferred
Medchemexpress, HY-10205 200mg Cediranib AZD2171 CAS:288383-20-0 Formula:C25H27FN4O3 IC50: <1 nM (Flt1), <3 nM (KDR), 5 nM (Flt4), 5 nM (PDGFRα), 36 nM (PDGFRβ), 2 nM (c-Kit) Purity:98% Cediranib maleate (AZD-2171 maleate) is a highly potent, orally available VEGFR inhibitor with IC 50 s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2577 Supplier Medchemexpress LLC Supplier No. HY10205200MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-10205 200mg Cediranib AZD2171 CAS:288383-20-0 Formula:C25H27FN4O3 IC50: <1 nM (Flt1), <3 nM (KDR), 5 nM (Flt4), 5 nM (PDGFRα), 36 nM (PDGFRβ), 2 nM (c-Kit) Purity:98% Cediranib maleate (AZD-2171 maleate) is a highly potent, orally available VEGFR inhibitor with IC 50 s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.