Learn More
Apexbio Technology LLC Cediranib (AZD217) 288383-20-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063097
Encompass_Preferred
Cediranib (AZD2171 CAS 288383-20-0) is an orally bioavailable small molecule inhibitor targeting the kinase insert domain receptor (KDR VEGFR-2) with potent inhibition observed in recombinant assays (IC50 1 nM) Additionally cediranib suppresses related vascular endothelial growth factor receptors VEGFR-1 (Flt-1 IC50 5 nM) and VEGFR-3 (Flt-4 IC50 3 nM) It also blocks select platelet-derived growth factor receptors notably c-Kit PDGFR PDGFR and CSF-1R at nanomolar to sub-micromolar concentrations indicating multiple-kinase targeting capability Cediranib serves as a valuable research compound for studying tumor angiogenesis and kinase-driven oncogenic pathways
Catalog No. 50-000-63097 Supplier Apexbio Technology LLC Supplier No. A18825.1
Add to Cart
Add to Cart
Cediranib (AZD2171 CAS 288383-20-0) is an orally bioavailable small molecule inhibitor targeting the kinase insert domain receptor (KDR VEGFR-2) with potent inhibition observed in recombinant assays (IC50 1 nM) Additionally cediranib suppresses related vascular endothelial growth factor receptors VEGFR-1 (Flt-1 IC50 5 nM) and VEGFR-3 (Flt-4 IC50 3 nM) It also blocks select platelet-derived growth factor receptors notably c-Kit PDGFR PDGFR and CSF-1R at nanomolar to sub-micromolar concentrations indicating multiple-kinase targeting capability Cediranib serves as a valuable research compound for studying tumor angiogenesis and kinase-driven oncogenic pathways