Learn More
Apexbio Technology LLC Cediranib (AZD217), 5mg. Cas: 288383-20-0 MFCD: MFCD09954115. VEGFR inhibitor receptor,highly potent
SDP
Supplier:  Apexbio Technology LLC A18825
Encompass_Preferred
Cediranib (also known as AZD2171) is a highly potent KDR tyrosine kinase inhibitor that ATP-competitively inhibits recombinant KDR tyrosine kinase as well as other members of vascular endothelial growth factor receptor (VEGFR) family Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4716