Learn More
Selleck Chemical LLC Cediranib Maleate S5242-10mg
SDP
Catalog No. 503111997
Encompass_Preferred
Cediranib Maleate (AZD2171) is the maleate salt of Cediranib which is a potent inhibitor of VEGFR with IC50 of 1 nM and also inhibits Flt1/4 with IC50 of 5 nM/ 3 nM
Catalog No. 50-311-1997 Supplier Selleck Chemical LLC Supplier No. S524210MG
Cediranib Maleate (AZD2171) is the maleate salt of Cediranib which is a potent inhibitor of VEGFR with IC50 of 1 nM and also inhibits Flt1/4 with IC50 of 5 nM/ 3 nM