Learn More
Medchemexpress LLC HY-13049 5mg Medchemexpress, Cediranib (maleate) CAS:857036-77-2 Purity:>98%
SDP
Catalog No. 502081242
Encompass_Preferred
Medchemexpress, HY-13049 5mg Cediranib (maleate) CAS:857036-77-2 Cediranib maleate (AZD-2171 maleate) is a highly potent, orally available VEGFR inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1242 Supplier Medchemexpress LLC Supplier No. HY130495MG
Medchemexpress, HY-13049 5mg Cediranib (maleate) CAS:857036-77-2 Cediranib maleate (AZD-2171 maleate) is a highly potent, orally available VEGFR inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.